Product Details

SNP ID
rs13184275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10568961 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGGGTGTTTCCCTTTACTCTTGA[C/G]GTGCTTAAAATTAAAACTTAATTTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD33B PubMed Links

Gene Details

Gene
ANKRD33B
Gene Name
ankyrin repeat domain 33B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164440.1 Intron NP_001157912.1
XM_005248343.4 Intron XP_005248400.1
XM_005248344.3 Intron XP_005248401.1

View Full Product Details