Product Details

SNP ID
rs11264403
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:155706701 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAATCGCTTGAGCCCGGGGAGGCA[A/G]GGTTCCAGTGAACTCCAGCTTGGGT
Phenotype
MIM: 602074
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAP3 PubMed Links

Gene Details

Gene
DAP3
Gene Name
death associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199849.1 Intron NP_001186778.1
NM_001199850.1 Intron NP_001186779.1
NM_001199851.1 Intron NP_001186780.1
NM_004632.3 Intron NP_004623.1
NM_033657.2 Intron NP_387506.1
XM_005245480.2 Intron XP_005245537.1
XM_005245481.2 Intron XP_005245538.1
XM_005245482.4 Intron XP_005245539.1
XM_011509972.2 Intron XP_011508274.1
XM_017002289.1 Intron XP_016857778.1
XM_017002290.1 Intron XP_016857779.1
XM_017002291.1 Intron XP_016857780.1
XM_017002292.1 Intron XP_016857781.1
XM_017002293.1 Intron XP_016857782.1
XM_017002294.1 Intron XP_016857783.1
XM_017002295.1 Intron XP_016857784.1
XM_017002296.1 Intron XP_016857785.1
XM_017002297.1 Intron XP_016857786.1

View Full Product Details