Product Details
- SNP ID
-
rs12206362
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:7331153 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCTTTCTCTCTTTCTTTTTTTAC[A/G]TCTTTAATGAAATGACCTTGGAAAT
- Phenotype
-
MIM: 608304
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAGE1
PubMed Links
Gene Details
- Gene
- CAGE1
- Gene Name
- cancer antigen 1
View Full Product Details