Product Details

SNP ID
rs12664567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:13624348 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCTCCCTATCTAAACTACTCTTT[A/C]ATCTACTTTTTGTGGCACTGCCCTC
Phenotype
MIM: 611533 MIM: 603854 MIM: 604483
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NOL7 PubMed Links

Gene Details

Gene
NOL7
Gene Name
nucleolar protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317724.1 Intron NP_001304653.1
NM_016167.4 Intron NP_057251.2
Gene
RANBP9
Gene Name
RAN binding protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005493.2 Intron NP_005484.2
XM_006714945.2 Intron XP_006715008.1
XM_011514205.2 Intron XP_011512507.1
XM_017010149.1 Intron XP_016865638.1
Gene
SIRT5
Gene Name
sirtuin 5
There are no transcripts associated with this gene.

View Full Product Details