Product Details

SNP ID
rs11169559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50789454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATCTTTATAATGTAAGTGAACC[A/C]GTGGACAGAGAAGTAGTATGAAGTT
Phenotype
MIM: 123803
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATF1 PubMed Links

Gene Details

Gene
ATF1
Gene Name
activating transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005171.4 Intron NP_005162.1
XM_011538386.2 Intron XP_011536688.1
XM_011538387.2 Intron XP_011536689.1
XM_011538388.2 Intron XP_011536690.1
XM_017019331.1 Intron XP_016874820.1
XM_017019332.1 Intron XP_016874821.1
XM_017019333.1 Intron XP_016874822.1
XM_017019334.1 Intron XP_016874823.1
XM_017019335.1 Intron XP_016874824.1
XM_017019336.1 Intron XP_016874825.1

View Full Product Details