Product Details

SNP ID
rs10758347
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36137372 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACACCTTCCTCACCATTGTCCTC[A/G]GAGTGAAAGGACACACTCCAGTCCT
Phenotype
MIM: 607141
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GLIPR2 PubMed Links
Additional Information
For this assay, SNP(s) [rs77847577] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GLIPR2
Gene Name
GLI pathogenesis related 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287010.1 Intron NP_001273939.1
NM_001287011.1 Intron NP_001273940.1
NM_001287012.1 Intron NP_001273941.1
NM_001287013.1 Intron NP_001273942.1
NM_001287014.1 Intron NP_001273943.1
NM_022343.3 Intron NP_071738.1

View Full Product Details