Product Details

SNP ID
rs12376826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101391814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAAGAAAGACTGGAATTACATT[C/T]TACACCAAATGGACTTGACACTTAG
Phenotype
MIM: 602938 MIM: 611854 MIM: 603132
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAAT PubMed Links

Gene Details

Gene
BAAT
Gene Name
bile acid-CoA:amino acid N-acyltransferase
There are no transcripts associated with this gene.

Gene
MRPL50
Gene Name
mitochondrial ribosomal protein L50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019051.2 Intron NP_061924.1
Gene
ZNF189
Gene Name
zinc finger protein 189
There are no transcripts associated with this gene.

View Full Product Details