Product Details

SNP ID
rs10980965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111606616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAATTGAGAGCCTCCCTGTGGTT[C/T]TAATCCACTTGGGGCAGAATCTCTC
Phenotype
MIM: 601274
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC37A5P PubMed Links

Gene Details

Gene
LRRC37A5P
Gene Name
leucine rich repeat containing 37 member A5, pseudogene
There are no transcripts associated with this gene.

Gene
PTGR1
Gene Name
prostaglandin reductase 1
There are no transcripts associated with this gene.

View Full Product Details