Product Details
- SNP ID
-
rs11591839
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:127878938 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGAACTGGGTACGGAGACTGGGCT[G/T]CCCTGAGAGCCACAAATGCCATCCC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLRN3
PubMed Links
Gene Details
- Gene
- CLRN3
- Gene Name
- clarin 3
View Full Product Details