Product Details

SNP ID
rs11190326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99930658 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTAGAGTCCATCCAGCACTGTAA[A/G]GTTCCTTTTCCGTACACTCCCGTAG
Phenotype
MIM: 611282
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNMBP PubMed Links

Gene Details

Gene
DNMBP
Gene Name
dynamin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318326.1 339 Missense Mutation CTT,TTT L36F NP_001305255.1
NM_001318327.1 339 Intron NP_001305256.1
NM_015221.3 339 Intron NP_056036.1
XM_006717735.2 339 Intron XP_006717798.1
XM_011539559.2 339 Intron XP_011537861.1
Gene
DNMBP-AS1
Gene Name
DNMBP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details