Product Details

SNP ID
rs13288863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:76458168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATATCACAGTCTAATGGCCTCT[C/G]AGTTGGATTTTTTTTTTTTTTTTTT
Phenotype
MIM: 600391
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GCNT1 PubMed Links

Gene Details

Gene
GCNT1
Gene Name
glucosaminyl (N-acetyl) transferase 1, core 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097633.1 Intron NP_001091102.1
NM_001097634.1 Intron NP_001091103.1
NM_001097635.1 Intron NP_001091104.1
NM_001097636.1 Intron NP_001091105.1
NM_001490.4 Intron NP_001481.2
XM_017014620.1 Intron XP_016870109.1
XM_017014621.1 Intron XP_016870110.1

View Full Product Details