Product Details

SNP ID
rs10761237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:93578535 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGACCTGGCGAGCGCAGTCACTC[C/T]GCTACAGAACCTTCTGGGCACCTGT
Phenotype
MIM: 604351
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHF2 PubMed Links

Gene Details

Gene
PHF2
Gene Name
PHD finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005392.3 Intron NP_005383.3
XM_005252051.2 Intron XP_005252108.1
XM_006717143.2 Intron XP_006717206.1

View Full Product Details