Product Details

SNP ID
rs10750625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102051202 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTCAGTTTCAGGCATGCTGAATT[C/T]GAGGTACATACAAGTGGCAGTATTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf70 PubMed Links

Gene Details

Gene
C11orf70
Gene Name
chromosome 11 open reading frame 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195005.1 Intron NP_001181934.1
NM_032930.2 Intron NP_116319.2
XM_005271713.3 Intron XP_005271770.1
XM_017018454.1 Intron XP_016873943.1

View Full Product Details