Product Details

SNP ID
rs11225303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102522816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTTAACCGCTTTCTCTACATGTA[C/T]GTGCAAACAGGAAGAGCAGGGTCAC
Phenotype
MIM: 178990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP7 PubMed Links

Gene Details

Gene
MMP7
Gene Name
matrix metallopeptidase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002423.4 Intron NP_002414.1

View Full Product Details