Product Details

SNP ID
rs12294712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:110183111 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAAGCTTGCTAATGTGGGTTACC[C/T]GCACCTTGGTTCAACCACCAATGAA
Phenotype
MIM: 179410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RDX PubMed Links

Gene Details

Gene
RDX
Gene Name
radixin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260492.1 Intron NP_001247421.1
NM_001260493.1 Intron NP_001247422.1
NM_001260494.1 Intron NP_001247423.1
NM_001260495.1 Intron NP_001247424.1
NM_001260496.1 Intron NP_001247425.1
NM_002906.3 Intron NP_002897.1

View Full Product Details