Product Details

SNP ID
rs11219737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124612437 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACATTTCCCAAACAGGTGTGCCAG[C/T]CTCCCACCGCCTCTTTATTCATCCA
Phenotype
MIM: 608422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PANX3 PubMed Links

Gene Details

Gene
PANX3
Gene Name
pannexin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052959.2 Intron NP_443191.1

View Full Product Details