Product Details

SNP ID
rs11219906
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125112303 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTTAATTCTTCCATTAGCCTTG[C/T]GAAACAGACAAAGGCAAAGATCATT
Phenotype
MIM: 613066
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KRT18P59 PubMed Links

Gene Details

Gene
KRT18P59
Gene Name
keratin 18 pseudogene 59
There are no transcripts associated with this gene.

Gene
PKNOX2
Gene Name
PBX/knotted 1 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022062.2 307 Intron NP_071345.2
XM_005271642.2 307 Intron XP_005271699.1
XM_005271643.2 307 Intron XP_005271700.1
XM_006718894.2 307 Intron XP_006718957.1
XM_011542944.2 307 Intron XP_011541246.1
XM_011542945.2 307 Intron XP_011541247.1
XM_011542946.1 307 Intron XP_011541248.1
XM_011542947.2 307 Intron XP_011541249.1
XM_017018110.1 307 UTR 5 XP_016873599.1
XM_017018111.1 307 Intron XP_016873600.1
XM_017018112.1 307 Intron XP_016873601.1
Gene
TMEM218
Gene Name
transmembrane protein 218
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080546.2 307 Intron NP_001074015.1
NM_001258238.1 307 Intron NP_001245167.1
NM_001258239.1 307 Intron NP_001245168.1
NM_001258240.1 307 Intron NP_001245169.1
NM_001258241.1 307 Intron NP_001245170.1
NM_001258242.1 307 Intron NP_001245171.1
NM_001258243.1 307 Intron NP_001245172.1
NM_001258244.1 307 Intron NP_001245173.1
NM_001258245.1 307 Intron NP_001245174.1
NM_001258246.1 307 Intron NP_001245175.1
NM_001258247.1 307 Intron NP_001245176.1
XM_006718779.3 307 Intron XP_006718842.1
XM_006718780.3 307 Intron XP_006718843.1
XM_006718781.3 307 Intron XP_006718844.1
XM_006718783.3 307 Intron XP_006718846.1
XM_017017322.1 307 Intron XP_016872811.1
XM_017017323.1 307 Intron XP_016872812.1
XM_017017324.1 307 Intron XP_016872813.1

View Full Product Details