Product Details

SNP ID
rs12279684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5806087 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAGCCTTCTAAAGGAAGCGTAT[C/G]GTTGGTAGAGGTAGCAGCATTAATA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR52N2 PubMed Links

Gene Details

Gene
OR52N2
Gene Name
olfactory receptor family 52 subfamily N member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005174.1 2655 Intron NP_001005174.1
XM_011520090.2 2655 UTR 5 XP_011518392.1

View Full Product Details