Product Details

SNP ID
rs13409084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223052969 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCTGGTTGTCATGTCCTTCTAC[A/G]GCATTTTCTTGATCGGAATCATGCT
Phenotype
MIM: 607775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNE4 PubMed Links

Gene Details

Gene
KCNE4
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080671.3 507 Missense Mutation AGC,GGC S98G NP_542402.3

View Full Product Details