Product Details

SNP ID
rs8104138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52572266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGGTTTCTCCATGTTGGTCAG[G/T]ATGGTCTCGAACTCCCGACATCAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF701 PubMed Links

Gene Details

Gene
ZNF701
Gene Name
zinc finger protein 701
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172655.1 250 Missense Mutation GAT,TAT D9Y NP_001166126.1
NM_018260.2 250 Intron NP_060730.2
XM_011527092.1 250 Intron XP_011525394.1
Gene
ZNF808
Gene Name
zinc finger protein 808
There are no transcripts associated with this gene.

View Full Product Details