Product Details

SNP ID
rs11230311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60430292 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAACCCGAAGAGATGGAGAGTC[C/T]TCTGGGCCTGAGAGTGTGACAATAC
Phenotype
MIM: 606499
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MS4A5 PubMed Links

Gene Details

Gene
MS4A5
Gene Name
membrane spanning 4-domains A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023945.2 Intron NP_076434.2

View Full Product Details