Product Details

SNP ID
rs9333578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65122296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGCAGGCAGCTCGCGCAGGAC[C/G]GGGCGGGACCAGACAGTTGCGCGCA
Phenotype
MIM: 134690 MIM: 606866 MIM: 608046 MIM: 603414 MIM: 604575
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAU PubMed Links

Gene Details

Gene
FAU
Gene Name
FAU, ubiquitin like and ribosomal protein S30 fusion
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001997.4 Intron NP_001988.1
Gene
MIR6751
Gene Name
microRNA 6751
There are no transcripts associated with this gene.

Gene
MRPL49
Gene Name
mitochondrial ribosomal protein L49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004927.3 Intron NP_004918.1
Gene
SYVN1
Gene Name
synoviolin 1
There are no transcripts associated with this gene.

Gene
TM7SF2
Gene Name
transmembrane 7 superfamily member 2
There are no transcripts associated with this gene.

Gene
ZNHIT2
Gene Name
zinc finger HIT-type containing 2
There are no transcripts associated with this gene.

View Full Product Details