Product Details

SNP ID
rs13241786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100889035 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACATAGGCTTCCGTGCCTGACCC[G/T]ACGCAGACTCCCAGCAAGGCCTTGG
Phenotype
MIM: 100740 MIM: 614469 MIM: 611481
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACHE PubMed Links

Gene Details

Gene
ACHE
Gene Name
acetylcholinesterase (Cartwright blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000665.4 684 Intron NP_000656.1
NM_001282449.1 684 Intron NP_001269378.1
NM_001302621.1 684 Intron NP_001289550.1
NM_001302622.1 684 Intron NP_001289551.1
NM_015831.2 684 Intron NP_056646.1
XM_006715995.2 684 Intron XP_006716058.1
XM_011516225.1 684 Intron XP_011514527.1
XM_011516226.1 684 Intron XP_011514528.1
XM_011516228.2 684 Intron XP_011514530.1
XM_011516229.2 684 Intron XP_011514531.1
XM_017012219.1 684 Intron XP_016867708.1
XM_017012220.1 684 Intron XP_016867709.1
Gene
SRRT
Gene Name
serrate, RNA effector molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128852.1 684 Intron NP_001122324.1
NM_001128853.1 684 Intron NP_001122325.1
NM_001128854.1 684 Intron NP_001122326.1
NM_015908.5 684 Intron NP_056992.4
XM_005250405.2 684 Intron XP_005250462.1
XM_005250406.2 684 Intron XP_005250463.1
XM_005250407.2 684 Intron XP_005250464.1
XM_005250408.2 684 Intron XP_005250465.1
XM_017012290.1 684 Intron XP_016867779.1
XM_017012291.1 684 Intron XP_016867780.1
Gene
UFSP1
Gene Name
UFM1 specific peptidase 1 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015072.3 684 Silent Mutation GTA,GTC V79V NP_001015072.2

View Full Product Details