Product Details

SNP ID
rs12974008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11090620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAGGGGAGGCCTGGCGTTCCTC[C/G]GCGGTTCCTGTCACAAAGGCGACGA
Phenotype
MIM: 606945
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LDLR PubMed Links

Gene Details

Gene
LDLR
Gene Name
low density lipoprotein receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000527.4 Intron NP_000518.1
NM_001195798.1 Intron NP_001182727.1
NM_001195799.1 Intron NP_001182728.1
NM_001195800.1 Intron NP_001182729.1
NM_001195803.1 Intron NP_001182732.1
XM_011528010.1 Intron XP_011526312.1

View Full Product Details