Product Details

SNP ID
rs11879567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20925940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGTGTAAAAGAAGTGGGTTTTA[C/G]AAAAAAATTAAAATCTAATCATATA
Phenotype
MIM: 603899
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF85 PubMed Links

Gene Details

Gene
ZNF85
Gene Name
zinc finger protein 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256171.1 Intron NP_001243100.1
NM_001256172.1 Intron NP_001243101.1
NM_001256173.1 Intron NP_001243102.1
NM_003429.4 Intron NP_003420.2
XM_011528263.2 Intron XP_011526565.1
XM_011528264.1 Intron XP_011526566.1
XM_017027234.1 Intron XP_016882723.1

View Full Product Details