Product Details

SNP ID
rs10981696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113229137 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGGATTATAGGCGTGATCCACTG[C/T]GCCTGACTGATAATTTAATTTTTTA
Phenotype
MIM: 603085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP15 PubMed Links

Gene Details

Gene
FKBP15
Gene Name
FK506 binding protein 15
There are no transcripts associated with this gene.

Gene
SLC31A1
Gene Name
solute carrier family 31 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001859.3 Intron NP_001850.1

View Full Product Details