Product Details

SNP ID
rs10818778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:123027472 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCCTGGAGGCCCAGCCACATTTT[G/T]CTAGTCTGGAAAAAATAATGTATTC
Phenotype
MIM: 601909 MIM: 615882
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR21 PubMed Links

Gene Details

Gene
GPR21
Gene Name
G protein-coupled receptor 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005294.2 131 Intron NP_005285.1
XM_005251933.3 131 UTR 5 XP_005251990.1
Gene
RABGAP1
Gene Name
RAB GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012197.3 131 Intron NP_036329.3
XM_011518440.2 131 Intron XP_011516742.1
XM_011518441.2 131 Intron XP_011516743.1
XM_011518442.2 131 Intron XP_011516744.1
XM_011518444.2 131 Intron XP_011516746.1
XM_017014567.1 131 Intron XP_016870056.1
XM_017014568.1 131 Intron XP_016870057.1
XM_017014569.1 131 Intron XP_016870058.1
XM_017014570.1 131 Intron XP_016870059.1

View Full Product Details