Product Details

SNP ID
rs12376093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122271603 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACCTCTTAGACTAGGTCAAGTCC[A/C]CTGTTGCCTGCACTCGTGCATTCTG
Phenotype
MIM: 604602
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRRF PubMed Links

Gene Details

Gene
MRRF
Gene Name
mitochondrial ribosome recycling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173512.1 Intron NP_001166983.1
NM_138777.3 Intron NP_620132.1
NM_199177.2 Intron NP_954646.1
XM_005252302.3 Intron XP_005252359.1
XM_011519183.2 Intron XP_011517485.1
XM_011519184.2 Intron XP_011517486.1
XM_011519185.2 Intron XP_011517487.1
XM_011519186.1 Intron XP_011517488.1
XM_011519187.2 Intron XP_011517489.1
XM_017015286.1 Intron XP_016870775.1
XM_017015287.1 Intron XP_016870776.1
Gene
RBM18
Gene Name
RNA binding motif protein 18
There are no transcripts associated with this gene.

View Full Product Details