Product Details
- SNP ID
-
rs11568204
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:6208306 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAAGTTTCCAGTGACACCTCTGAG[C/T]AGGCTAAGCAGCTGGTGGCTCCTGC
- Phenotype
-
MIM: 143030
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD9
PubMed Links
Gene Details
- Gene
- CD9
- Gene Name
- CD9 molecule
View Full Product Details