Product Details

SNP ID
rs6976847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27129437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGCGCTCAGACAAACAGAGCG[G/T]GTGGGCGATCTCGATGCGGCGCCGC
Phenotype
MIM: 616068 MIM: 142954 MIM: 142953
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HOXA-AS2 PubMed Links

Gene Details

Gene
HOXA-AS2
Gene Name
HOXA cluster antisense RNA 2
There are no transcripts associated with this gene.

Gene
HOXA3
Gene Name
homeobox A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030661.4 798 Intron NP_109377.1
NM_153631.2 798 Intron NP_705895.1
XM_005249730.2 798 Intron XP_005249787.1
XM_005249731.2 798 Intron XP_005249788.1
XM_005249732.3 798 Intron XP_005249789.1
XM_006715715.2 798 Intron XP_006715778.1
XM_011515343.2 798 Intron XP_011513645.1
Gene
HOXA4
Gene Name
homeobox A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002141.4 798 Missense Mutation ACG,CCG T251P NP_002132.3

View Full Product Details