Product Details

SNP ID
rs11685630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127541828 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCCAGGGCACCCTGCCTTCGCTG[A/T]CTTCCCCAGGAGGGGCTGCACCAGG
Phenotype
MIM: 606541
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MYO7B PubMed Links

Gene Details

Gene
MYO7B
Gene Name
myosin VIIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080527.1 Intron NP_001073996.1
XM_006712539.3 Intron XP_006712602.1
XM_011511218.2 Intron XP_011509520.1
XM_017004169.1 Intron XP_016859658.1

View Full Product Details