Product Details
- SNP ID
-
rs5744041
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:12905436 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATACCCACCCAAGCCATTCTTTGA[C/T]TGCTGACAGGCCTTCCTTCATAACA
- Phenotype
-
MIM: 300366
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TLR8
PubMed Links
Gene Details
- Gene
- TLR8
- Gene Name
- toll like receptor 8
There are no transcripts associated with this gene.
- Gene
- TLR8-AS1
- Gene Name
- TLR8 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details