Product Details

SNP ID
rs12134102
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:61683296 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAAAGGATTGGAGATGTTTCAGG[A/C]ATAGGGGTACCTGAGAGGTTTATAC
Phenotype
MIM: 610080
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TM2D1 PubMed Links

Gene Details

Gene
TM2D1
Gene Name
TM2 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032027.2 Intron NP_114416.1

View Full Product Details