Product Details

SNP ID
rs6943341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55907489 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTTTGTGGCTGCATGGTATTCC[A/T]CGTGCAGGTTTGTTACATAGACATG
Phenotype
MIM: 616181
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF713 PubMed Links

Gene Details

Gene
ZNF713
Gene Name
zinc finger protein 713
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182633.2 Intron NP_872439.2

View Full Product Details