Product Details

SNP ID
rs13197637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:124962872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACTGCAGTTGGAGCTGGAGGAG[G/T]AAGAGGAGGAAGCTGGGGATCGAAA
Phenotype
MIM: 602532
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RNF217 PubMed Links

Gene Details

Gene
RNF217
Gene Name
ring finger protein 217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286398.2 463 Missense Mutation GAA,TAA E110* NP_001273327.1
NM_152553.4 463 Intron NP_689766.1
XM_011535494.2 463 Missense Mutation GAA,TAA E110* XP_011533796.1
XM_011535495.2 463 Nonsense Mutation GAA,TAA E110* XP_011533797.1
XM_011535496.2 463 Missense Mutation GAA,TAA E110* XP_011533798.1
XM_011535499.2 463 Missense Mutation GAA,TAA E110* XP_011533801.1
XM_011535500.2 463 Intron XP_011533802.1
XM_017010317.1 463 Missense Mutation GAA,TAA E110* XP_016865806.1
Gene
RNF217-AS1
Gene Name
RNF217 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details