Product Details

SNP ID
rs11702238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:14274383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAACTTGAACCAGAGGAGCTCA[G/T]TCTTGGTAGGGCTGGAGGAGAGGAA
Phenotype
MIM: 608835
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCC13 PubMed Links

Gene Details

Gene
ABCC13
Gene Name
ATP binding cassette subfamily C member 13 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details