Product Details
- SNP ID
-
hCV32277555
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:183119963 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCTTAAAGAGATGCTTCTACCCT[C/T]CCACCATGAGAGATTCTGATTTTTT
- Phenotype
-
MIM: 608140
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUP35
PubMed Links
Gene Details
- Gene
- NUP35
- Gene Name
- nucleoporin 35
There are no transcripts associated with this gene.
View Full Product Details