Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001281736.1 | 3148 | UTR 3 | NP_001268665.1 | ||
NM_006375.3 | 3148 | UTR 3 | NP_006366.2 | ||
NM_182314.2 | 3148 | UTR 3 | NP_872114.1 | ||
XM_005262354.3 | 3148 | UTR 3 | XP_005262411.1 | ||
XM_011531244.2 | 3148 | UTR 3 | XP_011529546.1 | ||
XM_011531245.2 | 3148 | UTR 3 | XP_011529547.1 | ||
XM_011531247.2 | 3148 | UTR 3 | XP_011529549.1 | ||
XM_011531249.2 | 3148 | UTR 3 | XP_011529551.1 | ||
XM_011531251.2 | 3148 | UTR 3 | XP_011529553.1 | ||
XM_017029226.1 | 3148 | UTR 3 | XP_016884715.1 | ||
XM_017029227.1 | 3148 | UTR 3 | XP_016884716.1 | ||
XM_017029228.1 | 3148 | UTR 3 | XP_016884717.1 | ||
XM_017029229.1 | 3148 | UTR 3 | XP_016884718.1 |