Product Details

SNP ID
rs12835421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120614550 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCATATTATTTATTCCTGGTCCAC[A/G]AAAGTATAAATAGTCTCATCAGGAA
Phenotype
MIM: 300587
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCTS1 PubMed Links

Gene Details

Gene
MCTS1
Gene Name
MCTS1, re-initiation and release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137554.1 2992 UTR 3 NP_001131026.1
NM_014060.2 2992 UTR 3 NP_054779.1

View Full Product Details