Product Details

SNP ID
rs4757429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:16742895 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCCACCTCTTTGGCAAATCTTG[C/T]ATACTCAAGTTCTGCAGCCAGCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf58 PubMed Links

Gene Details

Gene
C11orf58
Gene Name
chromosome 11 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014267.5 1728 Intron NP_055082.1
XM_017017143.1 1728 UTR 5 XP_016872632.1

View Full Product Details