Product Details

SNP ID
rs147474393
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:207322526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGAGAAGGACTCAGTGATCTGCC[G/T]TAAGGGCAGTCAATGGTCAGATATT
Phenotype
MIM: 125240
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD55 PubMed Links

Gene Details

Gene
CD55
Gene Name
CD55 molecule (Cromer blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000574.4 373 Missense Mutation CGT,CTT R82L NP_000565.1
NM_001114752.2 373 Missense Mutation CGT,CTT R82L NP_001108224.1
NM_001300902.1 373 Missense Mutation CGT,CTT R82L NP_001287831.1
NM_001300903.1 373 Missense Mutation CGT,CTT R82L NP_001287832.1
NM_001300904.1 373 Missense Mutation CGT,CTT R82L NP_001287833.1
XM_017000467.1 373 Missense Mutation CGT,CTT R82L XP_016855956.1

View Full Product Details