Product Details
- SNP ID
-
hCV32309852
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:40752789 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAAATGTCTCACTGATTATTTAG[A/C]CAGGAATATATATTTTTGGTTGTCT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NSUN7
PubMed Links
Gene Details
- Gene
- NSUN7
- Gene Name
- NOP2/Sun RNA methyltransferase family member 7
There are no transcripts associated with this gene.
View Full Product Details