Product Details

SNP ID
rs4329703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125906165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGGAAGTTTTACAGAAGGATCC[C/G]AGCTCTCCACTTTACTCAGTAAAGA
Phenotype
MIM: 607663 MIM: 610693 MIM: 616283
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX25 PubMed Links

Gene Details

Gene
DDX25
Gene Name
DEAD-box helicase 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013264.4 419 Silent Mutation CCC,CCG P89P NP_037396.3
Gene
HYLS1
Gene Name
HYLS1, centriolar and ciliogenesis associated
There are no transcripts associated with this gene.

Gene
PUS3
Gene Name
pseudouridylate synthase 3
There are no transcripts associated with this gene.

View Full Product Details