Product Details

SNP ID
rs4719605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:20136754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATGTGGGAGAAAAGTTCATACA[C/G]AAGGAGGAGGGACTTACTTCTCCTC
Phenotype
MIM: 612646
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927668 PubMed Links

Gene Details

Gene
LOC101927668
Gene Name
uncharacterized LOC101927668
There are no transcripts associated with this gene.

Gene
MACC1
Gene Name
metastasis associated in colon cancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182762.3 7060 UTR 3 NP_877439.3
Gene
MACC1-AS1
Gene Name
MACC1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details