Product Details

SNP ID
rs8027534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45635705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTATTTATCAAAATTCCAACC[C/T]GGCTTTTTGCTCAAAGACGCTGAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SQRDL PubMed Links

Gene Details

Gene
SQRDL
Gene Name
sulfide quinone reductase-like (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271213.1 Intron NP_001258142.1
NM_021199.3 Intron NP_067022.1

View Full Product Details