Product Details

SNP ID
rs4963956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:26121258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTTAGAAGAAATTAAGATGAGT[C/T]CATTAATTCATGGTCAATTTCTCAA
Phenotype
MIM: 606200 MIM: 601599
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BHLHE41 PubMed Links

Gene Details

Gene
BHLHE41
Gene Name
basic helix-loop-helix family member e41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030762.2 2548 UTR 3 NP_110389.1
Gene
SSPN
Gene Name
sarcospan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135823.1 2548 Intron NP_001129295.1
NM_005086.4 2548 Intron NP_005077.2
XM_011520853.2 2548 Intron XP_011519155.1
XM_011520854.2 2548 Intron XP_011519156.1
XM_011520855.2 2548 Intron XP_011519157.1

View Full Product Details