Product Details

SNP ID
rs1053920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10392569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGGTGAGTTCTTTTACTCGTCG[C/G]TCATGTTTCCGTAAACCTTTAACAG
Phenotype
MIM: 160741
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYH8 PubMed Links

Gene Details

Gene
MYH8
Gene Name
myosin heavy chain 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002472.2 5636 Missense Mutation GAC,GAG D1847E NP_002463.2
Gene
MYHAS
Gene Name
myosin heavy chain gene cluster antisense RNA
There are no transcripts associated with this gene.

View Full Product Details