Product Details

SNP ID
rs3747296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103063430 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTCACGTTTCCCACCAGAGAGG[C/T]TTCGTGCGCCCACTAGGGGGCCCCC
Phenotype
MIM: 300690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BEX1 PubMed Links

Gene Details

Gene
BEX1
Gene Name
brain expressed X-linked 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018476.3 Intron NP_060946.3

View Full Product Details