Product Details

SNP ID
rs3174500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103063250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTTCCATGCTGAGACTGTTTA[C/A]TGCTCGTTTCTCTTTGGACTCCATT
Phenotype
MIM: 300690
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
BEX1 PubMed Links

Gene Details

Gene
BEX1
Gene Name
brain expressed X-linked 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018476.3 265 Missense Mutation GTA,TTA V9L NP_060946.3

View Full Product Details