Product Details

SNP ID
rs3192187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:65512646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAAGACATTCAAAATTCCCCTG[T/G]GGTGGACAACTGAGTTGATGTGGCT
Phenotype
MIM: 300964 MIM: 300889
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
LAS1L PubMed Links

Gene Details

Gene
LAS1L
Gene Name
LAS1 like, ribosome biogenesis factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170649.1 2356 UTR 3 NP_001164120.1
NM_001170650.1 2356 UTR 3 NP_001164121.1
NM_031206.4 2356 UTR 3 NP_112483.1
XM_005262301.1 2356 UTR 3 XP_005262358.1
XM_005262304.1 2356 Intron XP_005262361.1
XM_005262305.3 2356 Intron XP_005262362.1
XM_005262306.3 2356 Intron XP_005262363.1
XM_011531045.1 2356 UTR 3 XP_011529347.1
XM_011531046.1 2356 Intron XP_011529348.1
XM_017029877.1 2356 UTR 3 XP_016885366.1
XM_017029878.1 2356 Intron XP_016885367.1
XM_017029879.1 2356 UTR 3 XP_016885368.1
XM_017029880.1 2356 UTR 3 XP_016885369.1
Gene
ZC3H12B
Gene Name
zinc finger CCCH-type containing 12B
There are no transcripts associated with this gene.

View Full Product Details